missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetaldehyde, 95%, stabilized

Product Code. 10641875 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10641875 1 g 1g
10071772 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10641875 Supplier Thermo Scientific Acros Supplier No. 376070010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60656-87-3 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60656-87-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00191779
InChI Key NFNOAHXEQXMCGT-UHFFFAOYSA-N
Synonym 2-benzyloxy acetaldehyde, benzyloxyacetaldehyde, benzyloxy acetaldehyde, phenylmethoxy acetaldehyde, acetaldehyde, phenylmethoxy, acetaldehyde,2-phenylmethoxy, acmc-20ahpl, benzyloxy-acetaldehyde, 2-phenylmethoxyethanal
PubChem CID 108989
SMILES O=CCOCC1=CC=CC=C1

Specifications

Density 1.0690g/mL
Boiling Point 118°C to 120°C (13.0 mmHg)
Flash Point 113°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula C6H5CH2OCH2CHO
Quantity 1 g
Beilstein 06,III,1476
Specific Gravity 1.069
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform
IUPAC Name 2-phenylmethoxyacetaldehyde
Formula Weight 150.18
Percent Purity 95%
Chemical Name or Material Benzyloxyacetaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.