missing translation for 'onlineSavingsMsg'
Learn More

Benzylideneacetone, 98+%

Product Code. 11460004
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460004 250 g 250g
11450004 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460004 Supplier Thermo Scientific Alfa Aesar Supplier No. A13537.30
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122-57-6 | C10H10O | 146.19 g/mol

Benzylideneacetone is used chiefly as a scent in the manufacture of perfume, drugs & electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylideneacetone is used chiefly as a scent in the manufacture of perfume, drugs & electroplating.

Solubility
Soluble in alcohol, chloroform, diethyl ether.

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-57-6
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00008779
InChI Key BWHOZHOGCMHOBV-BQYQJAHWSA-N
Synonym benzalacetone, benzylideneacetone, methyl styryl ketone, 4-phenylbut-3-en-2-one, acetocinnamone, 4-phenyl-3-buten-2-one, e-4-phenylbut-3-en-2-one, benzylidene acetone, trans-benzalacetone, benzalaceton
PubChem CID 637759
ChEBI CHEBI:78399
SMILES CC(=O)\C=C\C1=CC=CC=C1

Specifications

Melting Point 38°C to 42°C
Density 1.038
Boiling Point 261°C
Flash Point 116°C (241°F)
Odor Characteristic
Quantity 250 g
Beilstein 742046
Sensitivity Light sensitive
Merck Index 14,1137
Solubility Information Soluble in alcohol,chloroform,diethyl ether.
IUPAC Name (E)-4-phenylbut-3-en-2-one
Formula Weight 146.19
Percent Purity ≥98%
Chemical Name or Material Benzylideneacetone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.