missing translation for 'onlineSavingsMsg'
Learn More

Benzylhydrazine Dihydrochloride, TRC

Product Code. 30334865
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334865 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334865 Supplier Toronto Research Chemicals Supplier No. TRCB5354231G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzylhydrazine Dihydrochloride
CAS 20570-96-1
Molecular Formula C7H10N2 . 2HCl
Quantity 1 g
Synonym 1-Benzylhydrazine Dihydrochloride, N-Benzylhydrazine Dihydrochloride
InChI Formula InChI=1S/C7H10N2.2ClH/c8-9-6-7-4-2-1-3-5-7;;/h1-5,9H,6,8H2;2*1H
SMILES Cl.Cl.NNCc1ccccc1
IUPAC Name benzylhydrazine;dihydrochloride
Molecular Weight (g/mol) 195.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.