missing translation for 'onlineSavingsMsg'
Learn More

Benzylhydrazine dihydrochloride, 97%

Product Code. 15461058
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461058 25 g 25g
15451058 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461058 Supplier Thermo Scientific Alfa Aesar Supplier No. H54916.14
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20570-96-1 | C7H12Cl2N2 | 195.087 g/mol

Benzylhydrazine dihydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylhydrazine dihydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20570-96-1
Molecular Formula C7H12Cl2N2
Molecular Weight (g/mol) 195.087
MDL Number MFCD00012921
InChI Key MSJHOJKVMMEMNX-UHFFFAOYSA-N
Synonym benzylhydrazine dihydrochloride, benzylhydrazine.2hcl, 1-benzylhydrazine dihydrochloride, benzyl-hydrazine dihydrochloride, hydrazine, phenylmethyl-, dihydrochloride, hydrazine, benzyl-, dihydrochloride, phenylmethyl hydrazine dihydrochloride, benzyl hydrazine di hcl, benzyl-hydrazine hydrochloride, benzylhydrazinedihydrochloride
PubChem CID 146540
IUPAC Name benzylhydrazine;dihydrochloride
SMILES C1=CC=C(C=C1)CNN.Cl.Cl

Specifications

Melting Point 143°C to 145°C
Linear Formula C6H5CH2NHNH2·2HCl
Quantity 25 g
UN Number UN2811
Beilstein 3688990
Solubility Information Insoluble in water.
Formula Weight 195.09
Percent Purity 97%
Chemical Name or Material Benzylhydrazine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.