missing translation for 'onlineSavingsMsg'
Learn More

Benzylhydrazine dihydrochloride, 97%

Product Code. 10165783 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10165783 100 g Glass bottle 100g
10182940 5 g Glass bottle 5g
10477280 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10165783 Supplier Thermo Scientific Acros Supplier No. 105961000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20570-96-1 | C7H10N2·2HCl | 195.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20570-96-1
Molecular Formula C7H10N2·2HCl
Molecular Weight (g/mol) 195.09
MDL Number MFCD00012921
InChI Key MSJHOJKVMMEMNX-UHFFFAOYSA-N
Synonym benzylhydrazine dihydrochloride, benzylhydrazine.2hcl, 1-benzylhydrazine dihydrochloride, benzyl-hydrazine dihydrochloride, hydrazine, phenylmethyl-, dihydrochloride, hydrazine, benzyl-, dihydrochloride, phenylmethyl hydrazine dihydrochloride, benzyl hydrazine di hcl, benzyl-hydrazine hydrochloride, benzylhydrazinedihydrochloride
PubChem CID 146540
IUPAC Name benzylhydrazine;dihydrochloride
SMILES C1=CC=C(C=C1)CNN.Cl.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 138°C to 143°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH2NHNH2·2HCl
Quantity 100 g
Beilstein 15,531
Formula Weight 195.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Benzylhydrazine dihydrochloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Benzylhydrazine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.