missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethylsilane, 96%

Product Code. 15418839
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418839 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15418839 Supplier Thermo Scientific Alfa Aesar Supplier No. L05334.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1631-70-5 | C9H13Si | 149.288 g/mol

Benzyldimethylsilane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyldimethylsilane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 69°C to 71°C (15mmHg)
CAS 1631-70-5
Quantity 5 g
Molecular Formula C9H13Si
MDL Number MFCD00039792
UN Number UN1993
Beilstein 2935572
Flash Point 53°C (127°F)
Synonym benzyldimethylsilane, dimethyl phenylmethyl silane, silane, dimethyl phenylmethyl, benzyldimethylsilyl, benzyl dimethyl silicon, benzene, dimethylsilyl methyl
Solubility Information Insoluble in water.
InChI Key SKDKWDVYZOKBPL-UHFFFAOYSA-N
SMILES C[Si](C)CC1=CC=CC=C1
IUPAC Name benzyl(dimethyl)silicon
Molecular Weight (g/mol) 149.288
PubChem CID 6328719
Formula Weight 150.3
Percent Purity 96%
Refractive Index 1.5026
Density 0.949
Chemical Name or Material Benzyldimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.