missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethylhexadecylammonium chloride hydrate, 97%

Product Code. 10626241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10626241 500 g Glass bottle 500g
10556972 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10626241 Supplier Thermo Scientific Acros Supplier No. 402075000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122-18-9 | C25H46ClN | 396.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 122-18-9
Molecular Formula C25H46ClN
Molecular Weight (g/mol) 396.10
MDL Number MFCD00149967
InChI Key SXPWTBGAZSPLHA-UHFFFAOYSA-M
Synonym cetalkonium chloride, benzyldimethylhexadecylammonium chloride, banicol, bonjela, cetyl zephiran, cetyldimethylbenzylammonium chloride, bicetonium, acinol, benzylhexadecyldimethylammonium chloride, baktonium
PubChem CID 31202
IUPAC Name benzyl-hexadecyl-dimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Melting Point 54.0°C to 58.0°C
Color Cream-White to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on anhydrous substance) (Argentometry)
Packaging Glass bottle
Quantity 500 g
Beilstein 12, III, 2212
Merck Index 15, 2016
Solubility Information Solubility in water: 8.5g/L. Other solubilities: soluble in alcohol, acetone, esters, ethyla acetat, e, propylene glycol, sorbitol solutions, glycerol,, ether
Formula Weight 396.1
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Benzyldimethylhexadecylammonium chloride hydrate, 97 - 101%
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Benzyldimethylhexadecylammonium chloride, hydrate
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
Gefahrenhinweis
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.