missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethylhexadecylammonium chloride, 95%

Product Code. 11378347
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378347 250 g 250g
11388347 10 g 10g
11398347 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378347 Supplier Thermo Scientific Alfa Aesar Supplier No. 032377.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-18-9 | C25H46ClN | 396.10 g/mol

Benzyldimethylhexadecylammonium chloride has been used in a study to assess the removal of cationic surfactants from aqueous solutions using activated carbon cloth (ACC). It has also been used in a study to investigate the prototropic behavior of the dye, 7-hydroxy-4-methylcoumarin (7H4MC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-18-9
Molecular Formula C25H46ClN
Molecular Weight (g/mol) 396.10
MDL Number MFCD00149967
InChI Key SXPWTBGAZSPLHA-UHFFFAOYSA-M
Synonym cetalkonium chloride, benzyldimethylhexadecylammonium chloride, banicol, bonjela, cetyl zephiran, cetyldimethylbenzylammonium chloride, bicetonium, acinol, benzylhexadecyldimethylammonium chloride, baktonium
PubChem CID 31202
IUPAC Name benzyl-hexadecyl-dimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Melting Point 54°C to 58°C
Color White
Linear Formula CH3(CH2)15N(CH2C6H5)(CH3)2Cl·xH2O
Quantity 250 g
UN Number UN3261
Beilstein 3802341
Sensitivity Hygroscopic
Merck Index 14,2017
Solubility Information Soluble in water (8.5g/L),ethanol,acetone,ethyl acetate,propylene glycol,sorbitol solutions,ether,glycerol,and carbon tetrachloride.
Formula Weight 396.1
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Benzyldimethylhexadecylammonium chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.