missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethyldodecylammonium Chloride, Mikromol™

Product Code. 17971716
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971716 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971716 Supplier Mikromol Supplier No. MM0224.01
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzyldimethyldodecylammonium Chloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyldimethyldodecylammonium Chloride
CAS 139-07-1
Analyte or Component Names Benzyldimethyldodecylammonium Chloride
Shipping Condition Room Temperature
Molecular Formula C21 H38 N . Cl
Certifications/Compliance ISO 17034
Synonym Benzenemethanaminium, N-dodecyl-N,N-dimethyl-, chloride (1:1), Ammonium, benzyldodecyldimethyl-, chloride (8CI), Benzenemethanaminium, N-dodecyl-N,N-dimethyl-, chloride (9CI), Benzyldodecyldimethylammonium chloride (6CI), Accelerator 1227, BAC 12, Bas 2631, Benzododecinium chloride, Benzyldimethyldodecylammonium chloride, Benzyldimethyllaurylammonium chloride, Benzyllauryldimethylammonium chloride, Catigene OM, Catinal CB 50, Catiogen BC 50, Catiogen PAN, Catiolite BC 50, Cationic 1227, Cequartyl A, Croda N-Dodecyl, DYK 1125, Dehyquart LDB, Dimethylaurylbenzylammonium chloride, Dimethylbenzyldodecylammonium chloride, Dimethylbenzyllaurylammonium chloride, Dimethyldodecylbenzylammonium chloride, Dimethyllaurylbenzylammonium chloride, Dodecylbenzyldimethylammonium chloride, Dodecyldimethylbenzylammonium chloride, Germicide 1227, Katamin AB 18, Lauralkonium chloride, Lauryl benzalkonium chloride, Laurylbenzyldimethylammonium chloride, Lauryldimethylbenzylammonium chloride, Leveling Agent 1227, Loraquat B 50, N-Benzyl-N-dodecyl-N,N-dimethylammonium chloride, N-Dodecyl-N,N-dimethyl-N-benzylammonium chloride, N-Dodecyl-N,N-dimethylbenzenemethanaminium chloride, N-Lauryl-N,N-dimethyl-N-benzylammonium chloride, Noramium DA 50, Orthosan HM, Pionin B 231, QBA 1211, Retarder N, Rewoquat B 50, Rolcril, Surfactant 1227, Swanol CA 100, Swanol CA 101, Tetranil BC 80, Texnol R 5, Vantoc CL, Zephirol
InChI Formula InChI=1S/C21H38N.ClH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCCCCCC[N+](C)(C)Cc1ccccc1
IUPAC Name benzyl-dodecyl-dimethylazanium;chloride
Molecular Weight (g/mol) 339.99
Quantity 500 mg
Formula Weight 339.2693 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.