missing translation for 'onlineSavingsMsg'
Learn More

Benzylbenzene, TRC

Product Code. 30336743
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30336743 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30336743

Brand: Toronto Research Chemicals TRCB287265100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Diphenylmethane
CAS 101-81-5
Molecular Formula C13 H12
Quantity 100 g
Synonym Methane, diphenyl- (8CI), 1,1'-Methylenebis[benzene], 1,1-Diphenylmethane, Benzene, (phenylmethyl)-, Benzylbenzene, DPM, Diphenylmethane, Ditan, Ditane, NSC 4708
InChI Formula InChI=1S/C13H12/c1-3-7-12(8-4-1)11-13-9-5-2-6-10-13/h1-10H,11H2
SMILES C(c1ccccc1)c2ccccc2
IUPAC Name benzylbenzene
Molecular Weight (g/mol) 168.23
Formula Weight 168.0939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.