missing translation for 'onlineSavingsMsg'
Learn More

Benzylamine hydrochloride, 99%, Thermo Scientific™

£12.20 - £77.70

Chemical Identifiers

CAS 3287-99-8
Molecular Formula C7H9N·ClH
Molecular Weight (g/mol) 143.62
MDL Number MFCD00012852
InChI Key XKXHCNPAFAXVRZ-UHFFFAOYSA-N
Synonym benzylamine hydrochloride, phenylmethanamine hydrochloride, benzenemethanamine, hydrochloride, benzylammonium chloride, benzylamine, hydrochloride, usaf el-82, benzenemethanamine, hydrochloride 1:1, benzylamine hcl, benzylaminehydrochloride, benzyl amine hcl salt
PubChem CID 2724127
IUPAC Name phenylmethanamine;hydrochloride
SMILES C1=CC=C(C=C1)CN.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10694712
SDS View Documents Promotion Details
Thermo Scientific Acros
200540050
5g Glass bottle
£12.20
5g
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10173830
SDS View Documents Promotion Details
Thermo Scientific Acros
200541000
100g Plastic bottle
£77.70
100g
View alternative products
Save Up to 
»
Estimated Shipment: 07-07-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3287-99-8
143.62
XKXHCNPAFAXVRZ-UHFFFAOYSA-N
2724127
C1=CC=C(C=C1)CN.Cl
C7H9N·ClH
MFCD00012852
benzylamine hydrochloride, phenylmethanamine hydrochloride, benzenemethanamine, hydrochloride, benzylammonium chloride, benzylamine, hydrochloride, usaf el-82, benzenemethanamine, hydrochloride 1:1, benzylamine hcl, benzylaminehydrochloride, benzyl amine hcl salt
phenylmethanamine;hydrochloride

Specifications

Benzylamine hydrochloride
3287-99-8
100.0
259°C to 264°C
Authentic
C6H5CH2NH2·HCl
12, 1017
14, 1125
5g
XKXHCNPAFAXVRZ-UHFFFAOYSA-N
phenylmethanamine;hydrochloride
2724127
Crystals or Crystalline Powder
99%
98.5
White
99%
C7H9N·ClH
MFCD00012852
Glass bottle
benzylamine hydrochloride, phenylmethanamine hydrochloride, benzenemethanamine, hydrochloride, benzylammonium chloride, benzylamine, hydrochloride, usaf el-82, benzenemethanamine, hydrochloride 1:1, benzylamine hcl, benzylaminehydrochloride, benzyl amine hcl salt
Solubility in water: soluble
C1=CC=C(C=C1)CN.Cl
143.62
143.62
99%
Safety and Handling

Safety and Handling

Product Identifier
  • Benzylamine hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Product Certifications

RUO – Research Use Only