missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Benzylamine hydrochloride, 99%

Product Code. 11386786
Change view
Click to view available options
Quantity:
100g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386786 100g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11386786 Supplier Alfa Aesar™ Supplier No. L04102.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3287-99-8
Molecular Formula C7H10ClN
MDL Number MFCD00012852
Quantity 100g
Beilstein 3594394
Merck Index 14,1125
Synonym benzylamine hydrochloride, phenylmethanamine hydrochloride, benzenemethanamine, hydrochloride, benzylammonium chloride, benzylamine, hydrochloride, usaf el-82, benzenemethanamine, hydrochloride 1:1, benzylamine hcl, benzylaminehydrochloride, benzyl amine hcl salt
InChI Key XKXHCNPAFAXVRZ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)CN.Cl
IUPAC Name phenylmethanamine;hydrochloride
Molecular Weight (g/mol) 143.614
PubChem CID 2724127
Formula Weight 143.62
Percent Purity 99%
Chemical Name or Material Benzylamine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.