missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylamine, 99%, pure, Thermo Scientific™
£19.60 - £712.40
Chemical Identifiers
CAS | 100-46-9 |
---|---|
Molecular Formula | C7H9N |
Molecular Weight (g/mol) | 107.15 |
MDL Number | MFCD00008106 |
InChI Key | WGQKYBSKWIADBV-UHFFFAOYSA-N |
Synonym | benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine |
PubChem CID | 7504 |
ChEBI | CHEBI:40538 |
IUPAC Name | phenylmethanamine |
SMILES | C1=CC=C(C=C1)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10203990
|
Thermo Scientific Acros
105851000 |
100ml | Glass bottle |
£19.60
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10795931
|
Thermo Scientific Acros
105855000 |
500ml | Glass bottle |
£90.80
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10338993
|
Thermo Scientific Acros
105850025 |
2.5l | Glass bottle |
£171.60
2.50 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11925241
|
Thermo Scientific Acros
105850100 |
10l | Plastic drum |
£712.40
10 L |
Estimated Shipment: 17-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
100-46-9 | |
107.15 | |
WGQKYBSKWIADBV-UHFFFAOYSA-N | |
7504 | |
phenylmethanamine |
C7H9N | |
MFCD00008106 | |
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
CHEBI:40538 | |
C1=CC=C(C=C1)CN |
Specifications
Benzylamine | |
100-46-9 | |
100.0 | |
Colorless to Light Yellow | |
182.0°C to 185.0°C | |
98.5% min. (GC) | |
C7H9N | |
MFCD00008106 | |
Glass bottle | |
15,1128 | |
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
Solubility in water: soluble in water. Other solubilities: >50g/100mL methanol,>50g/100mL ethanol,>50g/100mL 2-propanol,miscible with alcohol,ether,acetone,benzene | |
0.98 | |
phenylmethanamine | |
7504 | |
107.15 | |
99% | |
1.82 mPa.s (20°C) |
99% | |
98.5 | |
0.9800g/mL | |
-30.0°C | |
72°C | |
Authentic | |
C6H5CH2NH2 | |
12,1013 | |
01,51; 10,26; 13,79; 16,22 | |
1.5424 to 1.5444 | |
100ml | |
WGQKYBSKWIADBV-UHFFFAOYSA-N | |
C1=CC=C(C=C1)CN | |
107.15 | |
CHEBI:40538 | |
Liquid | |
Pure | |
0.3% max. (K.F.) |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Benzylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only