missing translation for 'onlineSavingsMsg'
Learn More

Benzylamine, 99%, pure

Product Code. 10338993 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
10 L
100mL
2.5 L
500mL
Product Code. Quantity Packaging unitSize
10338993 2.5 L Glass bottle 2.5 L
10203990 100 mL Glass bottle 100mL
10795931 500 mL Glass bottle 500mL
11925241 10 L Plastic drum 10 L
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10338993

Brand: Thermo Scientific Acros 105850025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 100-46-9 | C7H9N | 107.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-46-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.15
MDL Number MFCD00008106
InChI Key WGQKYBSKWIADBV-UHFFFAOYSA-N
Synonym benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
PubChem CID 7504
ChEBI CHEBI:40538
IUPAC Name phenylmethanamine
SMILES C1=CC=C(C=C1)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -30.0°C
Color Colorless to Light Yellow
Density 0.9800g/mL
Boiling Point 182.0°C to 185.0°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2NH2
Refractive Index 1.5424 to 1.5444
Quantity 2.5 L
Beilstein 12,1013
Fieser 01,51; 10,26; 13,79; 16,22
Merck Index 15,1128
Specific Gravity 0.98
Solubility Information Solubility in water: soluble in water. Other solubilities: >50g/100mL methanol,>50g/100mL ethanol,>50g/100mL 2-propanol,miscible with alcohol,ether,acetone,benzene
Viscosity 1.82 mPa.s (20°C)
Formula Weight 107.15
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Benzylamine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.