missing translation for 'onlineSavingsMsg'
Learn More

Benzylacetone, 99%

Product Code. 10020560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10020560 250 mL Glass bottle 250mL
10478240 5 mL Glass bottle 5mL
10474385 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10020560 Supplier Thermo Scientific Acros Supplier No. 105832500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2550-26-7 | C10H12O | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-26-7
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
MDL Number MFCD00008790
InChI Key AKGGYBADQZYZPD-UHFFFAOYSA-N
Synonym benzylacetone, 4-phenyl-2-butanone, 2-butanone, 4-phenyl, 4-penylbutan-2-one, 1-phenyl-3-butanone, methyl phenethyl ketone, benzyl acetone, methyl 2-phenylethyl ketone, phenethyl methyl ketone, methyl phenylethyl ketone
PubChem CID 17355
IUPAC Name 4-phenylbutan-2-one
SMILES CC(=O)CCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9875g/mL
Boiling Point 235°C to 237°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2COCH3
Refractive Index 1.5100 to 1.513
Quantity 250 mL
Beilstein 07, 314
Specific Gravity 0.9875
Solubility Information Solubility in water: practically insoluble in water.. Other solubilities: soluble in ethanol,diethyl ether,chloroform,,very soluble in acetone
Formula Weight 148.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzylacetone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Benzylacetone
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.