missing translation for 'onlineSavingsMsg'
Learn More

Benzyl viologen dichloride, 97%

Product Code. 15482829
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482829 5 g 5g
15472829 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15482829 Supplier Thermo Scientific Alfa Aesar Supplier No. H66836.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1102-19-8 | C24H22Cl2N2 | 409.354 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1102-19-8
Molecular Formula C24H22Cl2N2
Molecular Weight (g/mol) 409.354
MDL Number MFCD00011991
InChI Key NLOIIDFMYPFJKP-UHFFFAOYSA-L
Synonym 1,1'-dibenzyl-4,4'-bipyridinium dichloride, benzyl viologen, benzylviologen chloride, benzyl viologen dichloride, n,n'-dibenzyl-gamma,gamma'-dipyridylium dichloride, 1,1'-bis phenylmethyl-4,4'-bipyridinium dichloride, 4,4'-bipyridinium, 1,1'-dibenzyl-, dichloride, 4,4'-bipyridinium, 1,1'-bis phenylmethyl-, dichloride, 1,1'-dibenzyl-4,4'-bipyridine-1,1'-diium chloride, 1-benzyl-4-1-benzylpyridin-1-ium-4-yl pyridin-1-ium dichloride
PubChem CID 14195
IUPAC Name 1-benzyl-4-(1-benzylpyridin-1-ium-4-yl)pyridin-1-ium;dichloride
SMILES C1=CC=C(C=C1)C[N+]2=CC=C(C=C2)C3=CC=[N+](C=C3)CC4=CC=CC=C4.[Cl-].[Cl-]

Specifications

Melting Point 260°C (decomposition)
Color White
Quantity 5 g
Beilstein 3811585
Formula Weight 409.35
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Benzyl viologen dichloride
compliance-icons
Product Identifier
  • Benzyl viologen dichloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1,1'-Dibenzyl-4,4'-bipyridinium dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.