missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (triphenylphosphoranylidene)acetate, 97%

Product Code. 10176943 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10176943 100 g Glass bottle 100g
10077002 5 g Glass bottle 5g
10766383 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10176943 Supplier Thermo Scientific Acros Supplier No. 377221000
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 15097-38-8 | C27H23O2P | 410.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15097-38-8
Molecular Formula C27H23O2P
Molecular Weight (g/mol) 410.45
MDL Number MFCD00191787
InChI Key INKMLGJBBDRIQR-UHFFFAOYSA-N
Synonym benzyl 2-triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene-acetate, benzyl 2-triphenyl-??-phosphanylidene acetate, acetic acid, triphenylphosphoranylidene-, phenylmethyl ester, phenylmethyl 2-triphenylylidene acetate, benzyl 2-triphenyl, acmc-20ah9g, ksc526q3l
PubChem CID 3862746
IUPAC Name benzyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES O=C(OCC1=CC=CC=C1)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 120.0°C to 127.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)3P=CHCOOCH2C6H5
Quantity 100 g
Formula Weight 410.45
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Benzyl (triphenylphosphoranylidene)acetate
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl (triphenylphosphoranylidene)acetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.