missing translation for 'onlineSavingsMsg'
Learn More

Benzyl tert-butyl malonate, 95%

Product Code. 11378316
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378316 25 g 25g
11368316 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378316 Supplier Thermo Scientific Alfa Aesar Supplier No. L15533.14
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 72594-86-6 | C14H18O4 | 250.294 g/mol

Benzyl tert-butyl malonate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl tert-butyl malonate is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72594-86-6
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.294
MDL Number MFCD01075175
InChI Key XKXXXODAXXAFNP-UHFFFAOYSA-N
Synonym benzyl tert-butyl malonate, benzyltert-butylmalonate, tert-butyl benzyl malonate, 1-benzyl 3-tert-butyl propanedioate, tert-butyl 2-benzyloxycarbonyl acetate, propanedioic acid, 1,1-dimethylethyl phenylmethyl ester, tert-butyl phenylmethyl propane-1,3-dioate, pubchem3938, t-butyl benzyl malonate, ksc493q0n
PubChem CID 2736712
IUPAC Name 1-O-benzyl 3-O-tert-butyl propanedioate
SMILES CC(C)(C)OC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Refractive Index 1.486
Quantity 25 g
Beilstein 5434466
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 250.3
Percent Purity 95%
Chemical Name or Material Benzyl tert-butyl malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.