missing translation for 'onlineSavingsMsg'
Learn More

Benzyl sulfide, 98%, Thermo Scientific™

Product Code. 10603082 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10603082 5 g 5g
10440822 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10603082 Supplier Thermo Scientific Acros Supplier No. 335000050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 538-74-9
Molecular Formula C14H14S
Molecular Weight (g/mol) 214.33
MDL Number MFCD00004784
InChI Key LUFPJJNWMYZRQE-UHFFFAOYSA-N
Synonym dibenzyl sulfide, benzyl sulfide, dibenzyl sulphide, benzyl thioether, benzyl monosulfide, dibenzyl monosulfide, dibenzyl thioether, sulfide, dibenzyl, 1,3-diphenyl-2-thiapropane, benzene, 1,1'-thiobis methylene bis
PubChem CID 10867
IUPAC Name benzylsulfanylmethylbenzene
SMILES C(SCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 44°C to 49°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 1147
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether
Formula Weight 214.3
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Benzyl sulfide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.