missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (S)-(+)-glycidyl ether, 98+%

Product Code. 11308915
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308915 5 g 5g
11398905 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308915 Supplier Thermo Scientific Alfa Aesar Supplier No. L14033.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16495-13-9 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16495-13-9
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00054428
InChI Key QNYBOILAKBSWFG-SNVBAGLBSA-N
Synonym s-+-benzyl glycidyl ether, s-benzyloxymethyl-oxirane, benzyl s-+-glycidyl ether, s-2-benzyloxy methyl oxirane, 2s-2-benzyloxy methyl oxirane, s-o-benzylglycidol, s-benzyl glycidyl ether, +-benzyl glycidyl ether, s-benzyloxymethyl oxirane
PubChem CID 146296
SMILES C(OCC1=CC=CC=C1)[C@@H]1CO1

Specifications

Density 1.072
Boiling Point 130°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.517
Quantity 5 g
Beilstein 5246495
Optical Rotation +5.1° (c=5 in Toluene)
IUPAC Name (2S)-2-(phenylmethoxymethyl)oxirane
Formula Weight 164.2
Percent Purity ≥98%
Chemical Name or Material Benzyl (S)-(+)-glycidyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.