missing translation for 'onlineSavingsMsg'
Learn More

Benzyl propargyl ether, 97%

Product Code. 10026232 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10026232 5 g Glass bottle 5g
10165542 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10026232 Supplier Thermo Scientific Acros Supplier No. 380870050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4039-82-1 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4039-82-1
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00483987
InChI Key PAQVEXAFKDWGOT-UHFFFAOYSA-N
Synonym benzyl propargyl ether, benzylpropargyl ether, 1-benzyloxy-2-propyne, prop-2-yn-1-yloxy methyl benzene, benzene, 2-propynyloxy methyl, prop-2-ynyloxy methyl benzene, propargylbenzyl ether, benzyloxy prop-2-yne
PubChem CID 6917484
IUPAC Name prop-2-ynoxymethylbenzene
SMILES C#CCOCC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0110g/mL
Boiling Point 100°C to 101°C (18.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OCH2C∽CH
Refractive Index 1.5190 to 1.521
Quantity 5 g
Specific Gravity 1.011
Formula Weight 146.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzyl propargyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.