missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl N-(2,6-Dioxo-3-piperidyl)carbamate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Benzyl N-(2,6-Dioxo-3-piperidyl)carbamate
Specifications
Specifications
| Chemical Name or Material | Benzyl N-(2,6-Dioxo-3-piperidyl)carbamate |
| CAS | 24666-55-5 |
| Analyte or Component Names | Benzyl N-(2,6-Dioxo-3-piperidyl)carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H14 N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Carbamic acid, N-(2,6-dioxo-3-piperidinyl)-, phenylmethyl ester (ACI), 3-Piperidinecarbamic acid, 2,6-dioxo-, benzyl ester (6CI, 7CI, 8CI), Carbamic acid, (2,6-dioxo-3-piperidinyl)-, phenylmethyl ester (9CI), Phenylmethyl N-(2,6-dioxo-3-piperidinyl)carbamate (ACI), Benzyl (2,6-dioxopiperidin-3-yl)carbamate |
| InChI Formula | InChI=1S/C13H14N2O4/c16-11-7-6-10(12(17)15-11)14-13(18)19-8-9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H,14,18)(H,15,16,17) |
| SMILES | O=C(NC1CCC(=O)NC1=O)OCc2ccccc2 |
| IUPAC Name | benzyl N-(2,6-dioxopiperidin-3-yl)carbamate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction