missing translation for 'onlineSavingsMsg'
Learn More

Benzyl methyl ether, 97%

Product Code. 11415497
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415497 50 g 50g
11405497 10 g 10g
11425497 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415497 Supplier Thermo Scientific Alfa Aesar Supplier No. B23490.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 538-86-3 | C8H10O | 122.167 g/mol

Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-86-3
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00025901
InChI Key GQKZBCPTCWJTAS-UHFFFAOYSA-N
Synonym benzyl methyl ether, methoxymethyl benzene, methyl benzyl ether, benzene, methoxymethyl, ether, benzyl methyl, benzylmethylether, unii-f22rls78bd, .alpha.-methoxytoluene, f22rls78bd, methoxyphenylmethane
PubChem CID 10869
IUPAC Name methoxymethylbenzene
SMILES COCC1=CC=CC=C1

Specifications

Melting Point -53°C
Density 0.94
Boiling Point 173°C to 175°C
Flash Point 56°C (133°F)
Refractive Index 1.5015
Quantity 50 g
UN Number UN3271
Merck Index 14,1140
Solubility Information Insoluble in water.
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material Benzyl methyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.