missing translation for 'onlineSavingsMsg'
Learn More

Benzyl mercaptan, polymer supported, 1% cross-linked, 100-200 mesh, 2.0-4.0 mmol/g on poly(styrene-divinylbenzene), Thermo Scientific Chemicals

Catalog Number 11440790
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
100-53-8
C7H8S
124.20
MFCD00004867,MFCD00801588,MFCD01863867
UENWRTRMUIOCKN-UHFFFAOYSA-N
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
7509
phenylmethanethiol
SCC1=CC=CC=C1
This item is not returnable. View return policy
100-53-8
C7H8S
124.20
MFCD00004867,MFCD00801588,MFCD01863867
UENWRTRMUIOCKN-UHFFFAOYSA-N
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
7509
phenylmethanethiol
SCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Pink to White
Mesh Size 100 to 200 Mesh
Quantity 50 g
Physical Form Beads
Chemical Name or Material Benzyl mercaptan, Polymer supported, 1% cross-linked, 2.0 to 4.0 mmol/g on poly(styrene-divinylbenzene)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'