missing translation for 'onlineSavingsMsg'
Learn More

Benzyl mercaptan, 99%, Thermo Scientific Chemicals

Catalog Number 11490571
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100-53-8
C7H8S
124.20
MFCD00004867,MFCD00801588,MFCD01863867
UENWRTRMUIOCKN-UHFFFAOYSA-N
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
7509
phenylmethanethiol
SCC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100-53-8
C7H8S
124.20
MFCD00004867,MFCD00801588,MFCD01863867
UENWRTRMUIOCKN-UHFFFAOYSA-N
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
7509
phenylmethanethiol
SCC1=CC=CC=C1

A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point °C to 30°C
Density 1.053
Boiling Point 194°C to 195°C
Flash Point 70°C (158°F)
Assay Percent Range 0.99
Refractive Index 1.576
Quantity 100 g
UN Number UN2810
Beilstein 605864
Sensitivity Air Sensitive
Merck Index 14,9322
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 124.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl mercaptan
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'