missing translation for 'onlineSavingsMsg'
Learn More

gamma-Benzyl L-glutamate, 99%

Product Code. 10424345 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10424345 1 g Glass bottle 1g
10022820 5 g Glass bottle 5g
10157503 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10424345 Supplier Thermo Scientific Acros Supplier No. 225670010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1676-73-9 | C12H15NO4 | 237.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1676-73-9
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.25
MDL Number MFCD00002633
InChI Key BGGHCRNCRWQABU-JTQLQIEISA-N
Synonym h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
PubChem CID 122337
IUPAC Name (2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N

Specifications

Melting Point 162°C to 167°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2O2CCH2CH2CH(NH2)CO2H
Quantity 1 g
Specific Rotation Condition +27.20° (20°C c=2,1N HCL)
Specific Rotation + 27.2
Formula Weight 237.25
Percent Purity 99%
Physical Form Powder
Chemical Name or Material γ-Benzyl L-glutamate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.