missing translation for 'onlineSavingsMsg'
Learn More

Benzyl isovalerate, 98%, Thermo Scientific™

Product Code. 11496636
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11496636

Brand: Thermo Scientific Alfa Aesar B24852.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-38-8
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.26
MDL Number MFCD00048348
InChI Key HVJKZICIMIWFCP-UHFFFAOYSA-N
Synonym benzyl isovalerate, benzyl 3-methylbutyrate, benzyl isopentanoate, butanoic acid, 3-methyl-, phenylmethyl ester, isovaleric acid, benzyl ester, phenylmethyl 3-methylbutanoate, benzyl 3-methyl butyrate, benzyl isovalerianate, isopropyl acetic acid, benzyl ester, isopentanoic acid, phenylmethyl ester
PubChem CID 7651
IUPAC Name benzyl 3-methylbutanoate
SMILES CC(C)CC(=O)OCC1=CC=CC=C1

Specifications

Density 0.985
Boiling Point 245°C
Quantity 50 g
Formula Weight 192.26
Percent Purity 98%
Chemical Name or Material Benzyl isovalerate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.