missing translation for 'onlineSavingsMsg'
Learn More

Benzyl glycinate hydrochloride, 98%

Product Code. 11469370
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469370 5 g 5g
11459370 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469370 Supplier Thermo Scientific Alfa Aesar Supplier No. H50247.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2462-31-9 | C9H12ClNO2 | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2462-31-9
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00035442
InChI Key VLQHNAMRWPQWNK-UHFFFAOYSA-N
Synonym benzyl glycinate hydrochloride, benzyl 2-aminoacetate hydrochloride, glycine benzyl ester hydrochloride, h-dl-gly-obzl.hcl, benzyl glycinate hcl, h-gly-obzl.hcl, benzyl glycinate, h-gly-obzl hcl, glycine benzyl ester hcl, glycine, phenylmethyl ester, hydrochloride
PubChem CID 11701227
IUPAC Name benzyl 2-aminoacetate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)CN.Cl

Specifications

Melting Point 139°C to 140°C
Linear Formula H2NCH2CO2CH2C6H5·HCl
Quantity 5 g
Formula Weight 201.65
Percent Purity 98%
Chemical Name or Material Benzyl glycinate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.