missing translation for 'onlineSavingsMsg'
Learn More

Benzyl cyanide, 99+%

Product Code. 11965611 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 11965611

Brand: Thermo Scientific Acros 180940025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140-29-4 | C8H7N | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-29-4
Molecular Weight (g/mol) 117.15
InChI Key SUSQOBVLVYHIEX-UHFFFAOYSA-N
IUPAC Name 2-phenylacetonitrile
SMILES N#CCC1=CC=CC=C1

Specifications

Melting Point -24.0°C
Density 1.0100 g/mL
Boiling Point 233.0°C to 234.0°C
Flash Point 102°C
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C8H7N
Linear Formula C6H5CH2CN
Refractive Index 1.5220 to 1.5240
Quantity 2.5 L
Beilstein 09, 441
Merck Index 15, 1134
Specific Gravity 1.01
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight 117.15
Chemical Name or Material Benzyl cyanide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.