missing translation for 'onlineSavingsMsg'
Learn More

Benzyl cinnamate, 99%, Thermo Scientific Chemicals

Catalog Number 10607172
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
103-41-3
C16H14O2
238.29
MFCD00004789,MFCD00004789,MFCD00004789
NGHOLYJTSCBCGC-VAWYXSNFSA-N
benzyl alcohol cinnamic ester, benzylcinnamate, benzyl alcohol, cinnamate, wln: r1u1vo1r, benzyl .gamma.-phenylacrylate, benzyl cis-cinnamate, benzyl z-cinnamate, 2-propenoic acid, 3-phenyl-, phenylmethyl ester, z-benzyl 3-phenylacrylate, cinnamic acid, benzyl ester, z
15558051
benzyl (Z)-3-phenylprop-2-enoate
O=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1
This item is not returnable. View return policy
103-41-3
C16H14O2
238.29
MFCD00004789,MFCD00004789,MFCD00004789
NGHOLYJTSCBCGC-VAWYXSNFSA-N
benzyl alcohol cinnamic ester, benzylcinnamate, benzyl alcohol, cinnamate, wln: r1u1vo1r, benzyl .gamma.-phenylacrylate, benzyl cis-cinnamate, benzyl z-cinnamate, 2-propenoic acid, 3-phenyl-, phenylmethyl ester, z-benzyl 3-phenylacrylate, cinnamic acid, benzyl ester, z
15558051
benzyl (Z)-3-phenylprop-2-enoate
O=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 33°C to 37°C
Color Colorless to Yellow
Boiling Point 195°C to 200°C (5.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCO2CH2C6H5
Refractive Index 1.4025 to 1.4045
Quantity 500 g
Beilstein 09, 584
Merck Index 15, 1133
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether and oils,practically insoluble in propylene glycol and,glycerin
Formula Weight 238.29
Percent Purity 99%
Physical Form Crystalline Mass or Liquid After Melting
Chemical Name or Material Benzyl cinnamate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'