missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloromethyl ether, tech. 70%

Product Code. 15446688
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446688 50 mL 50mL
15456688 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446688 Supplier Thermo Scientific Alfa Aesar Supplier No. H60207.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3587-60-8 | C8H9ClO | 156.609 g/mol

Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.

Solubility
Soluble in water [2.7 g/L (25 C)]

Notes
Store at 2-8°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3587-60-8
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00000886
InChI Key LADPCMZCENPFGV-UHFFFAOYSA-N
Synonym benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl
PubChem CID 137983
IUPAC Name chloromethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCCl

Specifications

Density 1.126
Boiling Point 102°C
Linear Formula C6H5CH2OCH2Cl
Refractive Index 1.527
Quantity 50 mL
UN Number UN2810
Beilstein 1364034
Solubility Information Soluble in water [2.7g/L (25 C)]
Formula Weight 156.61
Percent Purity 70%
Grade Technical
Chemical Name or Material Benzyl chloromethyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.