missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloromethyl ether, 75%

Product Code. 10083892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10083892 10 g Glass bottle 10g
10310933 50 g Glass bottle 50g
10247852 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10083892 Supplier Thermo Scientific Acros Supplier No. 376950100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3587-60-8 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3587-60-8
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00000886
InChI Key LADPCMZCENPFGV-UHFFFAOYSA-N
Synonym benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl
PubChem CID 137983
IUPAC Name chloromethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCCl

Specifications

CAS Min % 70.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 102°C (14.0 mmHg)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 70% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OCH2Cl
Quantity 10 g
Specific Gravity 1.13
Formula Weight 156.61
Percent Purity 75%
Physical Form Liquid
Chemical Name or Material Benzyl chloromethyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzyl chloromethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.