missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Chloroformate, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl Chloroformate |
| CAS | 501-53-1 |
| Molecular Formula | C8H7ClO2 |
| Quantity | 50 g |
| Synonym | (Benzyloxy)carbonyl Chloride, Benzyl Chloridocarbonate, Benzyl Chlorocarbonate, Benzyl Chloroformate, Carbobenzoxy Chloride, Carbobenzyloxy Chloride, Chloroformic Acid Benzyl Ester, NSC 83466, Phenylmethyl Chloroformate, Cbz-Cl, CbzCl, Cbz chloride, Z-chloride |
| InChI Formula | InChI=1S/C8H7ClO2/c9-8(10)11-6-7-4-2-1-3-5-7/h1-5H,6H2 |
| SMILES | ClC(=O)OCc1ccccc1 |
| IUPAC Name | benzyl carbonochloridate |
| Molecular Weight (g/mol) | 170.59 |
| Formula Weight | 170.0135 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?