missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloroformate, 3M (50 wt%) solution in toluene

Product Code. 10265410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265410 500 mL Glass bottle 500mL
10773904 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10265410 Supplier Thermo Scientific Acros Supplier No. 124495000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 501-53-1 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl chloroformate
Name Note 50 wt. % Solution in Toluene
CAS 501-53-1, 108-88-3, 100-44-7
CAS Min % 45.0, 47.0
CAS Max % 53.0, 55.0
Color Pink to Yellow
Density 1.0100g/mL
Flash Point 7°C
Packaging Glass bottle
Molecular Formula C8H7ClO2
Linear Formula C6H5CH2OOCCl
MDL Number MFCD00000640
Quantity 500 mL
Beilstein 06, 437
Fieser 01,109; 02,59; 15,22
Merck Index 15, 1801
Specific Gravity 1.01
Synonym benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
Solubility Information Solubility in water: decomposes
InChI Key HSDAJNMJOMSNEV-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)COC(=O)Cl
IUPAC Name benzyl carbonochloridate
Molecular Weight (g/mol) 170.6
PubChem CID 10387
Formula Weight 170.6
Percent Purity 45 to 53 wt%
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.