missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl chloride, 99%, pure, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 1.1000g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass Bottle |
Molecular Formula | C7H7Cl |
Linear Formula | C6H5CH2Cl |
Refractive Index | 1.5370 to 1.5390 (20°C, 589nm) |
Show More |
Safety and Handling
Product Identifier
- Benzyl chloride, stabilized
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Germ cell mutagenicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'