missing translation for 'onlineSavingsMsg'
Learn More

Benzyl bromoacetate, 97%

Product Code. 11468267
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468267 250 g 250g
11458267 50 g 50g
11448267 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468267 Supplier Thermo Scientific Alfa Aesar Supplier No. A17630.30
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5437-45-6 | C9H9BrO2 | 229.073 g/mol

It is employed in biological research purpose. Benzyl bromoacetate was used in the alkylation of (-)-2,3-O-isopropylidene-D-threitol that afforded lipopeptide, 2-[(4R,5R)-5-({[(9H-fluoren-9-yl)methoxy]carbonylaminomethyl}-2,2-dimethyl-1,3-dioxolan-4-yl)methoxy]acetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in biological research purpose. Benzyl bromoacetate was used in the alkylation of (-)-2,3-O-isopropylidene-D-threitol that afforded lipopeptide, 2-[(4R,5R)-5-({[(9H-fluoren-9-yl)methoxy]carbonylaminomethyl}-2,2-dimethyl-1,3-dioxolan-4-yl)methoxy]acetic acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place. Store under 20˚C Freezer, Under Inert Atmosphere.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5437-45-6
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00000190
InChI Key JHVLLYQQQYIWKX-UHFFFAOYSA-N
Synonym benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate
PubChem CID 62576
IUPAC Name benzyl 2-bromoacetate
SMILES C1=CC=C(C=C1)COC(=O)CBr

Specifications

Density 1.46
Boiling Point 157°C to 159°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.544
Quantity 250 g
Beilstein 973658
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 229.08
Percent Purity 97%
Chemical Name or Material Benzyl bromoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.