missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Bromide, TRC

Product Code. 17980935
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980935 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980935 Supplier Toronto Research Chemicals Supplier No. TRCB23365025G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 100-39-0
Molecular Formula C7 H7 Br
Quantity 25 g
Synonym Benzene, (bromomethyl)-, Toluene, α-bromo- (8CI), (Bromomethyl)benzene, (Bromophenyl)methane, 1-(Bromomethyl)benzene, Benzyl bromide, NSC 8041, Phenylmethyl bromide, α-Bromotoluene, ω-Bromotoluene, alpha-Bromotoluene, (Bromomethyl)benzene, (Bromophenyl)methane, 1-(Bromomethyl)benzene, Benzyl Bromide, NSC 8041, Phenylmethyl Bromide, α-Bromotoluene, ω-Bromotoluene, (Bromomethyl)-benzene, BnBr
InChI Formula InChI=1S/C7H7Br/c8-6-7-4-2-1-3-5-7/h1-5H,6H2
SMILES BrCc1ccccc1
IUPAC Name bromomethylbenzene
Molecular Weight (g/mol) 171.03
Formula Weight 169.9731
Percent Purity >95%
Chemical Name or Material Benzyl Bromide
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.