missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl bromide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographic chemicals, synthetic resins, dyes and plasticizers and esters. It is also involved in the synthesis of (E)-(2-Iodovinyl)benzene by reacting it with diiodomethane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographi
Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographi
Specifications
Melting Point | -4°C |
Density | 1.438 |
Boiling Point | 198°C to 199°C |
Flash Point | 87°C (189°F) |
Odor | Pungent, Makes Eyes Water |
Refractive Index | 1.575 |
Quantity | 1000 g |
UN Number | UN1737 |
Beilstein | 385801 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Product Identifier
- Benzyl bromide
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308-IF exposed or concerned:.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'