missing translation for 'onlineSavingsMsg'
Learn More

Benzyl azide, 94%

Product Code. 11372878
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372878 50 g 50g
11392878 2 g 2g
11382878 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11372878 Supplier Thermo Scientific Alfa Aesar Supplier No. 032635.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-79-7 | C7H7N3 | 133.154 g/mol

Benzyl azide is used in the synthesis of 1,2,3-triazole derivatives. It serves as a reagent used to introduce a PhCH2N group into a molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-79-7
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD00013836
InChI Key UDLLFLQFQMACJB-UHFFFAOYSA-N
Synonym azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029
PubChem CID 12152
IUPAC Name azidomethylbenzene
SMILES C1=CC=C(C=C1)CN=[N+]=[N-]

Specifications

Color Brown to Colorless
Density 1.065
Boiling Point 82°C to 85°C (16 mmHg)
Flash Point 43°C (110°F)
Refractive Index 1.5341
Quantity 50 g
UN Number UN1993
Solubility Information Insoluble in waterMiscible with ethanol and diethyl ether; Immiscible with water.
Formula Weight 133.15
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Benzyl azide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.