missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, ReagentPlus™, ≥99%, Honeywell™

Product Code. 15690980 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
100mL
2.5 L
20 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15690980 1 L Glass Bottle 1 L
15680980 100 mL Glass Bottle 100mL
15620990 500 mL Glass Bottle 500mL
15600990 2.5 L Glass Bottle 2.5 L
15610990 20 L Stainless Steel Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15690980 Supplier Honeywell Research Chemicals Supplier No. 1080061L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Melting Point -15.4°C
Color Colorless
Density 1.045g/mL at 25°C
Boiling Point 205.3°C
Flash Point 96°C (204.8°F)
Ignition Point 817°F
Packaging Glass Bottle
Linear Formula C6H5CH2OH
Refractive Index n20/D 1.539(lit.)
Quantity 1 L
UN Number UN3334
Beilstein 878307
Vapor Density 3.7 (vs air)
Formula Weight 108.14g/mol
Vapor Pressure 3.75mmHg (77°C), 13.3mmHg (100°C)
Percent Purity ≥99%
Grade ReagentPlus
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.