missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BENZYL ALCOHOL PURISS., PH-QUALITY, Honeywell Riedel-de Haen™
Specifications
Specifications
| CAS | 100-51-6 |
| CAS Max % | ≤ 100% |
| Melting Point | -15.4°C |
| Color | Colorless |
| Density | 1.045 g/cm3 (20°C) |
| Boiling Point | 205.3°C (1013 hPa) |
| Flash Point | 100.4°C |
| Molecular Formula | C7H8O |
| Linear Formula | C6H5CH2OH |
| Quantity | 210 kg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?