missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, Mikromol™

Product Code. 17913266
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913266 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913266 Supplier Mikromol Supplier No. MM0795.000250
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzyl Alcohol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl alcohol
CAS 100-51-6
Analyte or Component Names Benzyl alcohol
Shipping Condition Room Temperature
Molecular Formula C7 H8 O
Certifications/Compliance ISO 17034
Synonym Benzalkonium Chloride Imp. A (EP), Benzyl Alcohol, Benzenemethanol, Benzyl alcohol (8CI), (Hydroxymethyl)benzene, Benzenecarbinol, Benzylic alcohol, NSC 8044, Phenylcarbinol, Phenylmethanol, Phenylmethyl alcohol, Sunmorl BK 20, TB 13G, alpha-Hydroxytoluene, alpha-Toluenol, Benzalkonium Chloride Impurity A, Benzalkonium Impurity A
InChI Formula InChI=1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2
SMILES OCc1ccccc1
IUPAC Name phenylmethanol
Molecular Weight (g/mol) 108.14
Quantity 250 mg
Formula Weight 108.0575 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.