missing translation for 'onlineSavingsMsg'
Learn More

Benzyl alcohol, For ACS analysis, Reagent grade, 99%, MP Biomedicals™

Product Code. 11436281 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436281 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11436281 Supplier MP Biomedicals Supplier No. 0219388280
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 100-51-6
Color Colorless
Density 1.0442g/cm3 at 22.5°C
Assay Percent Range 0.99
Molecular Formula C7H8O
MDL Number MFCD00004599,MFCD03792087
Quantity 100 mL
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
SMILES OCC1=CC=CC=C1
IUPAC Name phenylmethanol
Molecular Weight (g/mol) 108.14
PubChem CID 244
ChEBI CHEBI:17987
Formula Weight 108.13
Grade Reagent
Physical Form Liquid
Chemical Name or Material Benzyl alcohol, For ACS analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.