missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Acrylate (Stabilized with 50ppm of MEHQ), TRC

Product Code. 30348445
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348445 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348445 Supplier Toronto Research Chemicals Supplier No. TRCB1969755G
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl Acrylate
CAS 2495-35-4
Molecular Formula C10H10O2
Quantity 5 g
Synonym 2-Propenoic Acid Phenylmethyl Ester, Benzyl 2-Propenoate, Acrylic Acid Benzyl Ester, Fancryl FA-BZA, FA-BZA, NSC 20964, SR 432, Sartomer SR 432, V 160, V 180, Viscoat 160
InChI Formula InChI=1S/C10H10O2/c1-2-10(11)12-8-9-6-4-3-5-7-9/h2-7H,1,8H2
SMILES C=CC(=O)OCc1ccccc1
IUPAC Name benzyl prop-2-enoate
Molecular Weight (g/mol) 162.19
Formula Weight 162.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.