missing translation for 'onlineSavingsMsg'
Learn More

Benzyl acetoacetate, 97%

Product Code. 11452727
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452727 50 g 50g
11304589 25 g 25g
11314589 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452727 Supplier Thermo Scientific Alfa Aesar Supplier No. A14915.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5396-89-4 | C11H12O3 | 192.214 g/mol

Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

Solubility
It is soluble in alkali solutions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 5396-89-4
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00008785
InChI Key WOFAGNLBCJWEOE-UHFFFAOYSA-N
Synonym benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x
PubChem CID 142266
IUPAC Name benzyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OCC1=CC=CC=C1

Tekniske data

Density 1.113
Boiling Point 276°C
Flash Point 108°C (226°F)
Refractive Index 1.512
Quantity 50 g
Beilstein 2049615
Solubility Information It is soluble in alkali solutions.
Formula Weight 192.22
Percent Purity 97%
Chemical Name or Material Benzyl acetoacetate
compliance-icons
Produktidentifikator
  • Benzyl acetoacetate
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
Faresætning
  • H319-Causes serious eye irritation.
Sikkerhedssætning
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplerende oplysninger
  • MIXTURE LIST-Contain: Benzyl acetoacetate

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.