missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl acetate |
| CAS | 140-11-4 |
| Molecular Formula | C9 H10 O2 |
| Quantity | 10 g |
| Synonym | Acetic acid, benzyl ester (6CI,8CI), Benzyl alcohol, acetate (6CI), (Acetoxymethyl)benzene, Benzyl acetate, Benzyl ethanoate, NSC 4550, Phenylmethyl acetate, α-Acetoxytoluene |
| InChI Formula | InChI=1S/C9H10O2/c1-8(10)11-7-9-5-3-2-4-6-9/h2-6H,7H2,1H3 |
| SMILES | CC(=O)OCc1ccccc1 |
| IUPAC Name | benzyl acetate |
| Molecular Weight (g/mol) | 150.17 |
| Formula Weight | 150.0681 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?