missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Acetate, TRC

Product Code. 30336055
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336055 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336055 Supplier Toronto Research Chemicals Supplier No. TRCB22262550G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl acetate
CAS 140-11-4
Molecular Formula C9 H10 O2
Quantity 50 g
Synonym Acetic acid, benzyl ester (6CI,8CI), Benzyl alcohol, acetate (6CI), (Acetoxymethyl)benzene, Benzyl acetate, Benzyl ethanoate, NSC 4550, Phenylmethyl acetate, α-Acetoxytoluene
InChI Formula InChI=1S/C9H10O2/c1-8(10)11-7-9-5-3-2-4-6-9/h2-6H,7H2,1H3
SMILES CC(=O)OCc1ccccc1
IUPAC Name benzyl acetate
Molecular Weight (g/mol) 150.17
Formula Weight 150.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.