missing translation for 'onlineSavingsMsg'
Learn More

Benzyl acetate, 99%

Product Code. 11465467
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465467 500 g 500g
11475467 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465467 Supplier Thermo Scientific Alfa Aesar Supplier No. B23472.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140-11-4 | C9H10O2 | 150.177 g/mol

Benzyl acetate can be used as a high boiling point solvent in coatings, like ink coating, binding agent and paint remover. It is used in soap and other chemical essences and has the effect of promote in floral and fantasy essences in jasmine, white orchid, fragrant plantain lily, gekkakou, narcissus and other essences. It is a plasticizer for ionophore membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl acetate can be used as a high boiling point solvent in coatings, like ink coating, binding agent and paint remover. It is used in soap and other chemical essences and has the effect of promote in floral and fantasy essences in jasmine, white orchid, fragrant plantain lily, gekkakou, narcissus and other essences. It is a plasticizer for ionophore membranes.

Solubility
Soluble in water 3.1 g/L.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-11-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008712
InChI Key QUKGYYKBILRGFE-UHFFFAOYSA-N
Synonym acetic acid benzyl ester, benzyl ethanoate, acetic acid, phenylmethyl ester, phenylmethyl acetate, acetic acid, benzyl ester, alpha-acetoxytoluene, phenylmethyl ethanoate, acetoxymethyl benzene, benzylester kyseliny octove, caswell no. 081ea
PubChem CID 8785
ChEBI CHEBI:52051
IUPAC Name benzyl acetate
SMILES CC(=O)OCC1=CC=CC=C1

Specifications

Melting Point °C to 51°C
Density 1.053
Boiling Point 205°C to 207°C
Flash Point 95°C (203°F)
Assay Percent Range 99+%
Refractive Index 1.502
Quantity 500 g
Beilstein 1908121
Merck Index 14,1123
Solubility Information Soluble in water 3.1g/L.
Formula Weight 150.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.