missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 6-hydroxynicotinate, 99%, Thermo Scientific™

Product Code. 11494827
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11494827

Brand: Thermo Scientific Alfa Aesar B21807.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191157-01-4
Molecular Formula C13H11NO3
Molecular Weight (g/mol) 229.235
MDL Number MFCD02683072
InChI Key HGBIRINXNOIQIR-UHFFFAOYSA-N
Synonym benzyl 6-hydroxynicotinate, benzyl 6-hydroxypyridine-3-carboxylate, acmc-1byee, benzyl 6-hydroxy-nicotinate, 6-hydroxynicotinic acid benzyl ester, phenylmethyl 6-hydroxy-3-pyridinecarboxylate, phenylmethyl 6-hydroxypyridine-3-carboxylate, 6-oxo-1,6-dihydro-pyridine-3-carboxylic acid benzyl ester, 3-pyridinecarboxylicacid, 1,6-dihydro-6-oxo-, phenylmethyl ester
PubChem CID 7023014
IUPAC Name benzyl 6-oxo-1H-pyridine-3-carboxylate
SMILES C1=CC=C(C=C1)COC(=O)C2=CNC(=O)C=C2

Specifications

Melting Point 178°C to 181°C
Quantity 100 g
Formula Weight 229.24
Percent Purity 99%
Chemical Name or Material Benzyl 6-hydroxynicotinate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.