missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-Hydroxybenzoate (Benzyl Paraben), TRC

Product Code. 30005081
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005081 1 g 1g
30008265 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005081 Supplier Toronto Research Chemicals Supplier No. TRCB2885751G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-18-8
Molecular Formula C14 H12 O3
Molecular Weight (g/mol) 228.24
Synonym Benzoic acid, p-hydroxy-, benzyl ester (6CI,7CI,8CI), 4-(Benzyloxycarbonyl)phenol, 4-Hydroxybenzoic acid benzyl ester, 4-Hydroxybenzoic acid, phenylmethyl ester, Benzyl 4-hydroxybenzoate, Benzyl 4-hydroxylbenzoate, Benzyl Parasept, Benzyl Tegosept, Benzyl p-hydroxybenzoate, Benzylparaben, NSC 8080, Nipabenzyl, POB-BZ, Parosept, Solbrol Z, p-Hydroxybenzoic acid benzyl ester
IUPAC Name benzyl 4-hydroxybenzoate
SMILES Oc1ccc(cc1)C(=O)OCc2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H12O3/c15-13-8-6-12(7-9-13)14(16)17-10-11-4-2-1-3-5-11/h1-9,15H,10H2
Formula Weight 228.0786
Percent Purity >95%
Chemical Name or Material Benzyl 4-Hydroxybenzoate (Benzyl Paraben)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.